Calcium hypochlorite, tech., granular, 65% available chlorine
Calcium hypochlorite, tech., granular, 65% available chlorine
Calcium hypochlorite, tech., granular, 65% available chlorine
Acros Organics
Calcium hypochlorite, tech., granular, 65% available chlorine
CAS: 7778-54-3 | CaCl2O2 | 142.98 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3003400101 kgPlastic bottle
Catalog number 300340010
HSN 28281090
Price (SGD)
1,344.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
1,344.00
Each
Chemical Identifiers
CAS7778-54-3
IUPAC Namecalcium dihypochlorite
Molecular FormulaCaCl2O2
InChI KeyZKQDCIXGCQPQNV-UHFFFAOYSA-N
SMILES[Ca++].[O-]Cl.[O-]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Additional infoavailable chlorine: 72%
Additional infocalcium hydroxide (Ca(OH)2): 4.5%
Appearance (Form)Granules or granular solid
pH(1% aq. soln.) about 11.6
Additional infosodium chloride (NaCl): 8.5%
View more
Oxidation reagent
RUO – Research Use Only