Check Order Status
Pipette Pump™ Pipetters
Actual product may vary
Bel-Art
Pipette Pump™ Pipetters
Provides safe, accurate and trouble-free pipetting. Bel Art™ Pipette Pump™ Pipetters utilize an easy-to-use one-handed operation.
Have Questions?
Change view
Catalog Number
Color
Volume (Metric)
FIS13-683D
Red
25 mL
FIS13-683C
Green
10 mL
FKS11727854
Red
25 mL
3 Options
Catalog number
FIS13-683D
Price (SGD)
101.00
Add to cart
Color:
Red
Volume (Metric):
25 mL
Price (SGD)
101.00
Add to cart
Provides safe, accurate and trouble-free pipetting. Bel Art™ Pipette Pump™ Pipetters utilize an easy-to-use one-handed operation.
Pipettes fit smoothly into its flexible tapered chuck
Thumbwheel on side is rotated for precision aspirating or dispensing
Plunger may be pressed for quick emptying
Resistant to acids and alkalis
Disassemble all parts for cleaning and before autoclaving at 121ºC (250ºF) for 20 minutes; chuck should not be autoclaved
Replacement Silicone Rubber Chuck and Washers are available
Specifications
Adjustable Tip Spacing
No
Autoclavable
Autoclavable
Color
Red
Disposable
Reusable
For Use With
Pipette
Grip Type
Ergonomic
Material
Polypropylene
Model
Pipette Pump Pipettors
Number of Channels
1
Sterility
Non-sterile
Type
Pipette
Volume (Metric)
25 mL
For Use With (Equipment)
Pipets
Resistance
Acid and Alkali
Unit Size