Whiting & Davis™ Metal Mesh Gloves
Whiting & Davis™ Metal Mesh Gloves
Actual product may vary
Whiting & Davis™ Metal Mesh Gloves
Constructed of individually welded and joined stainless steel rings for maximum strength, flexibility and durability.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-069892X-Large
FIS19-069884Small
FIS19-069889Medium
FIS19-069891Large
Catalog number FIS19-069892
Price (SGD)
-
Size:
X-Large
  • Individually welded rings allow the mesh fabric to flow without harsh edges
  • Seams are 4-ring joined, sealing the seam so gaps are non-existent
  • Thumbs are attached with a 3-ring joint for a strong seam at a high wear area
  • Ergonomically designed and constructed with tapered fingers and palms to prevent sagging
  • Gloves are 3-step reversible to be worn on either hand
  • Mesh is manufactured following the European Standard of EN1082
Specifications
ColorSilver
HandAmbidextrous
MaterialStainless Steel Fiber
Product TypeCut-resistant Gloves
SizeX-Large
For Use With (Application)food service, knife and blade sharpening/cleaning, fishing, garment manufacturing, veterinary services, animal control, Paper and pulp industry
Glove Cuff StyleStrap with dome fastener
Unit Size