Stainless Steel Micro Spatula Reagent Scoop
Stainless Steel Micro Spatula Reagent Scoop
Actual product may vary
Stainless Steel Micro Spatula Reagent Scoop
Stainless steel micro spatula features a scoop on one end and flat edge on the other end.
Have Questions?
Catalog NumberLength (Metric)
FISS41700150 mm
Catalog number FISS41700
Price (SGD)
-
Length (Metric):
150 mm
  • Scoop on one end and flat edge on the other end
  • Ideal for adding small amounts of ingredients and mixing
  • Autoclavable and chemical resistant
  • Single Scoop
Specifications
Quantity1
AutoclavableAutoclavable
ShapeScoop End, Spatula End
ColorSilver
Product TypeDouble Ended Spatula
For Use With (Application)Measuring
Length (English)6 in.
Diameter (English) OuterHandle: 0.12 in.
Diameter (Metric) OuterHandle: 3 mm
Dimensions (L x W)Scoop: 0.51 x 0.20 in.; Flat Side: 1.96 x 0.27 in.
MaterialStainless Steel
Length (Metric)150 mm
Unit Size