DURAN™ Gas Washing Bottle, Drechsel type head
Actual product may vary
Fisherbrand
DURAN™ Gas Washing Bottle, Drechsel type head
Gas washing bottles are used for cleaning gases due to a flow through a suitable liquid.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
FKS10695591With filter disc
FKS10238490Without filter disc
Catalog number FKS10695591
Price (SGD)
402.00
Includes:
With filter disc
Price (SGD)
402.00
For gas filtration and cleaning, gas washing bottles are required which consist of a gas distribution tube, a suitable bottle and a gas outlet opening. In this test setup, the gas is forced to flow through a liquid before it is released. The gas distribution pipe is immersed in the liquid for this purpose, the gas interferes through it and impurities of the gas are bound in the liquid. The gas is cleaned or washed so to speak. A filter disk in the gas distribution tube creates air bubbles that are as small as possible and thus form as large a contact area as possible.
  • Drechsel type head
  • With screw-connection system
  • Fired-on, highly durable white print
  • Individual parts can also be ordered separately
  • Easy-to-read scale with graduation marks to estimate the content
Specifications
Capacity (Metric)500 mL
MaterialBorosilicate Glass (3.3)
TypeSize (Metric) Fitting 9 mm
IncludesWith filter disc
Porosity1
Standard Taper JointThread Size GL 45
Diameter (Metric) Inner Tubing9 mm
For Use With (Application)Cleaning (washing) of gases with solvents; Purification of gas mixtures; With screw thread for quick exchange of the liquid containing bottles; Filtering
Quantity1 Each
Unit Size