Manufactured from borosilicate glass for corrosion-free performance. Quickfit™ Borosilicate Glass Dreschel Bottles provide an inexpensive method for washing or drying gases. The robust, durable construction guards against accidental breakage.
Have Questions?
Catalog Number
Capacity (Metric)
Height (Metric)
Diameter (Metric) Outside
FKS11382055
125 mL
163 mm
Flat base: 102 mm
Catalog number FKS11382055
Price (SGD)
167.00
Capacity (Metric):
125 mL
Height (Metric):
163 mm
Diameter (Metric) Outside:
Flat base: 102 mm
Price (SGD)
167.00
Manufactured from borosilicate glass for corrosion-free performance. Quickfit™ Borosilicate Glass Dreschel Bottles provide an inexpensive method for washing or drying gases. The robust, durable construction guards against accidental breakage.