HDPE, Amber Carboy
Actual product may vary
Kimberly-Clark
HDPE, Amber Carboy
For Use With: Storing and dispensing solvents, buffers, reagents and powders in light sensitive applications; Closure Material: Polypropylene; Color: Amber
Have Questions?
Catalog NumberCapacity (Metric)
KLC1292110 L
Catalog number KLC12921
Price (SGD)
-
Capacity (Metric):
10 L
  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Material, volume, and metric graduation marks imprinted on carboys
Specifications
Capacity (English)2.6 gal.
Capacity (Metric)10 L
Closure MaterialPolypropylene
Closure Size83 mm
ColorAmber
Dimensions (L x W x H)273 x 163 x 615 mm
For Use With (Application)Storing and dispensing solvents, buffers, reagents and powders in light sensitive applications
Height (English)19.5 in.
Length (English)10.75 in.
MaterialHDPE
Product TypeRectangular Carboy
ShapeRectangular
Width (English)6.4 in.
Unit Size