Check Order Status
Eluent Solution, for Ion Chromatography, 1.7 mM NaHCO
3
- 1.8 mM Na
2
CO
3
, Ricca Chemical
Element Search
Structure Search
Eluent Solution, for Ion Chromatography, 1.7 mM NaHCO
3
- 1.8 mM Na
2
CO
3
, Ricca Chemical
For Ion Chromatography
Have Questions?
Catalog Number
Quantity
KLC28101
4 L
Catalog number
KLC28101
Price (SGD)
-
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS
7732-18-5, 497-19-8, 144-55-8
Specifications
Chemical Name or Material
Eluent Solution
Color
Colorless
Name Note
1.7mM NaHCO
CAS
7732-18-5
CAS Min %
98.0
View more