Check Order Status
Kimtech™ M3 Sterile Face Masks
Actual product may vary
Kimtech™ M3 Sterile Face Masks
Sterile masks feature non-irritating BiCoSof™ inner facing and bendable aluminum nosepiece. Available with earloops or soft ties.
Have Questions?
Change view
Catalog Number
Fastening Type
Includes
KLC62467
Ties
7 in. Soft Ties
KLC62470
Earloops
Knitted Earloops
FIS18-999-2405
Ties
7 in. Soft Ties
KLC62494
Ties
9 in. Soft Ties
4 Options
Catalog number
KLC62467
Price (SGD)
225.00
Add to cart
Fastening Type:
Ties
Includes:
7 in. Soft Ties
Price (SGD)
225.00
Add to cart
Sterile masks feature non-irritating BiCoSof™ inner facing and bendable aluminum nosepiece. Available with earloops or soft ties.
Gamma-sterilized to 10
-6
White with soft ties or earloops
Outer layer of apertured polyethylene film to provide protection from contamination
Polyester binding and polyester filter media provide bacteria filtration efficiency of 91.9% and particle filtration efficiency of 93.1%
Specifications
Product Type
Cleanroom Face Mask
Color
White
Fastening Type
Ties
Material
SMS
Size
Standard
Sterility
Sterile
Style
Pleated
Cleanroom Class
Class 1 (ISO 3)
Disposable
Single-use
Filter Efficiency
89.3% (PFE), 94.1% (BFE)
Includes
7 in. Soft Ties
Latex Free
Yes
Model
62467
Pore Size
0.1 μm (PFE), 3 μm (BFE)
Series
Kimtech™ M3 Sterile
Unit Size