SlimLine™ Cleanroom Mop Adapter
Actual product may vary
Micronova
SlimLine™ Cleanroom Mop Adapter
Flat head, cleanroom mop adapters that feature a unique 360° swivel joint that can be locked at 180°, for the ultimate ease and control of flat surface cleaning. Available electropolished stainless steel or polypropylene mop frame.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialProduct TypeLength (English)
MICQDSL-18Stainless Steel (304), Polyketone SwivelMop Adapter18 in.
MICQPSL-09PolypropyleneAdapter, ACME Threaded Handle Attachment9 in.
MICQPSL-18PolypropyleneMop Frame18 in.
MICQDSL-09Stainless Steel (304), Polyketone SwivelMop Adapter9 in.
MICQDSL-14Stainless Steel (304)Mop Adapter14 in.
Catalog number MICQDSL-18
Price (SGD)
549.00
Material:
Stainless Steel (304), Polyketone Swivel
Product Type:
Mop Adapter
Length (English):
18 in.
Price (SGD)
549.00
For floor and wall cleaning
The Micronova SlimLine™ Cleanroom Mop Adapters feature a unique 360° swivel joint that allows an easy glide for cleaning flat surfaces and can be locked at 180° for more controlled cleaning of walls and within hoods and chambers. Adapters are available with either an ACME threaded end for plastic handles or a Quick Disconnect attachment for stainless steel handles.
  • Flat head mop adapters with 360° plastic swivel joints
  • Joint can lock at 180° for more control
  • Available with 304 electropolished stainless steel or polypropylene frames
  • For use with the SlimLine, PadMop, SnapMop and PocketMop series mop covers
  • Adapters fit in the SlimLine and SlimT bucket systems
  • QDSL - stainless steel frame for use with SSU series stainless steel handles
  • QPSL - polypropylene frame for use with STA series plastic handles
  • QSSL - stainless steel frame for use with STA or ALUMEH series handles
  • Available from 9 to 18-inch lengths
  • Autoclavable
Specifications
Color CodeWhite
Dimensions4 x 18 in. (10 x 45.7 cm)
Product TypeMop Adapter
Head Type360 Swivel
ColorGray
Length (English)18 in.
SterilityNon-sterile
Length (Metric)45.7 cm
AutoclavableAutoclavable
Height (Metric)7.62 cm
Height (English)3 in.
Width (Metric)10.2 cm
Width (English)4 in.
Resistant ToChemicals
DisposableReusable
Quantity1 Ea.
Care InstructionsStore at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage
For Use With (Equipment)Snapmop or Pocketmop covers and SSU series handles
For Use With (Application)Cleaning and Sanitizing Cleanroom and Controlled Environments Floors, Walls
Certifications/ComplianceCertificate of Conformance, ISO 9001 Certified Facility
Color-codedNo
MaterialStainless Steel (304), Polyketone Swivel
Unit Size