Albumin, Bovine, Fatty Acid-Free
Albumin, Bovine, Fatty Acid-Free
Actual product may vary
MP Biomedicals
Albumin, Bovine, Fatty Acid-Free
Prepared by Fraction V method
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
MPB021524011010 g
MPB0215240180100 g
Catalog number MPB0215240110
Price (SGD)
363.00
Add to cart
Quantity:
10 g
Price (SGD)
363.00
Add to cart
  • Low fatty acids
  • Purity: >98%
  • pH: 7
For best cell culture performance, see alternate Mfr. No. 180575
Peters, T., Albumin:an overview and bibliography, Miles Laboratory, Research Products Division:Elkhart, IN. (1980).
Specifications
For Use With (Application)Immunohematology, Serology
Molecular Weight (g/mol)66.338
NameBovine Albumin
pH Range5.2 to 7
Quantity10 g
SourceBovine
Storage Requirements2° to 8°C