Check Order Status
Sulfamethoxazole, 100%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Sulfamethoxazole, 100%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0215671125
25 g
Catalog number
MPB0215671125
Price (SGD)
132.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
132.00
Add to cart
Specifications
Chemical Name or Material
Sulfamethoxazole
CAS
723-46-6
Color
White
Molecular Formula
C10H11N3O3S
Quantity
25 g
View more