Bovine Albumin, from New Zealand-sourced Bovine Plasma
Bovine Albumin, from New Zealand-sourced Bovine Plasma
Actual product may vary
MP Biomedicals
Bovine Albumin, from New Zealand-sourced Bovine Plasma
Have Questions?
Catalog NumberQuantity
MPB0218062080100 g
Catalog number MPB0218062080
Price (SGD)
825.00
Add to cart
Quantity:
100 g
Price (SGD)
825.00
Add to cart
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies
  • From New Zealand-sourced Bovine Plasma
  • Microbiological Grade
  • ≥97% Total Protein
  • Yellow to Brown Powder
  • Synonyms: BSA, Bovine albumin, Albumin from bovine serum
  • CAS Number: 9048-46-8
  • EC Number: 232-936-2
  • MDL Number: MFCD00130384
Specifications
Formulation≥97% Total Protein
NameBovine Albumin
Quantity100 g
Sourcefrom New Zealand-sourced Bovine Plasma
FormPowder
Purity or Quality GradeMicrobiological Grade
Unit Size