Check Order Status
β-Cyclodextrin, >98%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
β-Cyclodextrin, >98%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219005325
25 g
Catalog number
MPB0219005325
Price (SGD)
122.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
122.00
Add to cart
Specifications
Chemical Name or Material
β-Cyclodextrin
CAS
7585-39-9
Color
White
Assay Percent Range
>98%
Molecular Formula
C42H70O35
View more