▸
Organic compounds
▸
Organic Polymers
▸
Polyethylene Glycol, Off-White Powder, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Polyethylene Glycol, Off-White Powder, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219544501
1 kg
Catalog number
MPB0219544501
Price (SGD)
203.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
203.00
Add to cart
Specifications
Chemical Name or Material
Polyethylene Glycol
Color
White
CAS
25322-68-3
Physical Form
Powder
Recommended Storage
Store at room temperature.
View more
Molecular Biology, Immunology