Penicillin-Streptomycin (5000 IU/mL, 5 mg/mL)
Actual product may vary
MP Biomedicals
Penicillin-Streptomycin (5000 IU/mL, 5 mg/mL)
Dual antibiotic solution. Concentration: Penicillin 5000 IU/mL, Streptomycin 5 mg/mL
Have Questions?
Catalog NumberQuantity
MPB091670049100 mL
Catalog number MPB091670049
Price (SGD)
47.00
Quantity:
100 mL
Price (SGD)
47.00
The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Chemical Name or MaterialPenicillin-Streptomycin
Quantity100 mL
Unit Size