Ready to Use 5% 7X™ Solutions
Actual product may vary
MP Biomedicals
Ready to Use 5% 7X™ Solutions
A 1 gallon Ready to Use 5% solution of 7X that includes a hand-pump for convenient dispensing
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
MPB0976680937X; 1 gal.
MPB097668193ES 7X; 1 gal.
Catalog number MPB097668093
Price (SGD)
108.00
Description:
7X; 1 gal.
Price (SGD)
108.00
  • Ideal for glass and plastic labware
  • Contain powerful sequestering reagents to prevent deposits that could harm equipment or critical experimental work, and they will not etch delicate glassware or plastic labware
  • Environmentally friendly, phosphate-free ES formulation is completely soluble in water at any concentration and drains completely from glassware in seconds
Specifications
FormLiquid
Concentration7X
Description7X; 1 gal.
For Use With (Application)For Glass, Plastic, benchtop equipment and tubing
Volume (English)1 gal.
Unit Size
Contents & Storage
15°C to 30°C